Harris Industries™ Woven Plastic Barrier Tape

Extra-durable polypropylene tape

Manufacturer: HARRIS INDUSTRIES INC WB2YM

 View more versions of this product

Catalog No. 19-085285

  Check Availability
Add to Cart

    Specifications

    Length (English) 200 ft.
    For Use With (Application) Marking off-limit or hazardous areas and crowd control
    Length (Metric) 60.96m
    Tensile Strength 100 lb.
    Thickness 8mil
    Width (English) 2 in.
    Width (Metric) 5.08cm
    Color Yellow, Magenta
    Product Type Barrier Tape
    Material Polypropylene
    Dimensions (L x W) 200 ft. x 2 in. (60.96m x 5.08cm)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit