Learn More
Harris Industries Woven Plastic Barrier Tape
Extra-durable polypropylene tape
Manufacturer:  Harris Industries WB2RD
 View more versions of this product
Catalog No. 19-059418
Add to cart
Description
Description
Extra-durable polypropylene tape. Long-lasting solution for marking off-limits or hazardous areas. Also helpful for crowd control. Tensile strength is 100 lb. (45.4kg) for the 3/4 in. wide (1.9cm) tape and 300 lb. (136kg) for the 2 in. wide (5.1cm) tape.
Specifications
Specifications
200 ft. x 2 in. (60.96m x 5.08 cm)
Black, Red
Barrier Tape
Marking off-limit or hazardous areas and crowd control
100 lb.
2 in.
Safety
Polypropylene
200 ft.
60.96 m
8 mil
5.08 cm