Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HARS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112441
Description
HARS Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is a cytoplasmic enzyme which belongs to the class II family of aminoacyl-tRNA synthetases. The enzyme is responsible for the synthesis of histidyl-transfer RNA, which is essential for the incorporation of histidine into proteins. The gene is located in a head-to-head orientation with HARSL on chromosome five, where the homologous genes share a bidirectional promoter. The gene product is a frequent target of autoantibodies in the human autoimmune disease polymyositis/dermatomyositis.
Specifications
HARS | |
Polyclonal | |
Unconjugated | |
HARS1 | |
CMT2W; dead end homolog 1; Dnd1; FLJ20491; HARS; HARS1; HARSL; HARSL protein; HisRS; histidine translase; histidine tRNA ligase; histidine--tRNA ligase; histidine--tRNA ligase, cytoplasmic; Histidyl-tRNA synthetase; histidyl-tRNA synthetase 1; histidyl-tRNA synthetase, cytoplasmic; HRS; im:7136719; MMHRS; Unknown (protein for IMAGE:7951312); USH3B; zgc:92215 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3035 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12081 | |
HARS1 | |
Recombinant Human Histidine--tRNA ligase, cytoplasmic protein (1-180AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction