Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HARS2 Mouse anti-Human, Canine, Monkey, PE/Atto594, Clone: OTI5H2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271739PEATT594

Catalog No. N271739P594


Add to cart

Description

Description

HARS2 Monoclonal antibody specifically detects HARS2 in Human, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

HARS2
OTI5H2
Flow Cytometry
Affinity Purified, Conjugated
HARS2
Full length recombinant protein of human HARS2 (NP_036340) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
EC 6.1.1, EC 6.1.1.21, HARSLprobable histidyl-tRNA synthetase, mitochondrial, HARS-related, HARSRhistidine tRNA ligase 2, mitochondrial (putative), HisRS, histidine translase, Histidine--tRNA ligase, Histidine--tRNA ligase-like, histidyl-tRNA synthetase 2, mitochondrial (putative), histidyl-tRNA synthetase-like, HO3histidine-tRNA ligase homolog
Mouse
IgG2b
0.1 mL
Primary
23438.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit