Learn More
Harvard Apparatus™ LOOK™ Reverse Cutting 3/8 Circle Suture Needles
Feature cutting edge on the outer curve
Manufacturer:  Harvard Apparatus™ 510479
Catalog No. 14-516-134
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Designed to stay sharp to ensure continued passage through tissue
  • Attached to the suture by a swagging technique providing one continuous, even, smooth strand between the needle and suture material
  • Tempered to the right combination of hardness and tensile strength
  • Resists bending, catching, and breaking
  • Manufactured from top quality surgical stainless steel
  • Regular tests gauge needles sharpness and ductility
  • Individually packaged in sterile, peelable, double envelopes for maximum protection
  • Ideal for tough, difficult-to-penetrate tissues
Specifications
Specifications
Black
0.07 to 0.099mm
0.47 in.
C17
5 to 0
45cm
Circle Suture Needle
0.002 to 0.003 in.
For tough, difficult to penetrate tissues
12mm
1
17.7 in.
Stainless Steel
Each