Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harvard Apparatus™ Stronghold™ Lattice Rods

Sturdy rods made to metric specifications

Manufacturer:  HARVARD APPARATUS INC 53 2550

 View more versions of this product

Catalog No. 14-679-23

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

0.5 in.
Harvard Apparatus™ Stronghold™
75cm
Lattice Rod
1.3cm
29.5 in.
Stainless Steel
0.062 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit