Learn More
Thermo Scientific™ Harvey™ DI+ Cartridge System
Remove calcium, magnesium and other ionic impurities with the Thermo Scientific™ Harvey™ DI+ water cartridge system, complete with remote dispenser.
$1199.00
Specifications
Flow Rate 4L/min
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
13-998-290
View Documents
Thermo Scientific™
AY1273X4
Harvey DI+, Dispenser, Pura-lite and cartridge
Each for $1,199.00
Add to cart
Description
Description
Remove calcium, magnesium and other ionic impurities that can harm steam sterilizers with the Thermo Scientific™ Harvey™ DI+ Cartridge System. This turn-key system ships complete with a remote dispenser, Pura-Lite indicator and a high capacity, two-bed ion exchange cartridge. It is quick to set up, easy to use and requires only simple maintenance. Easily install the system with wall mount and eliminate the need to store containers of distilled water. The system accepts a broad range of other cartridges to remove impurities and produces pure water at a flow rater of 4L/min.
  • Light indicator notifies user when it is time to replace the cartridge.
  • Nuclear-grade resin is enclosed in a virgin polypropylene canister to purify tap water.
  • Simple maintenance: quarter-turn quick-release canister is easy to replace.
  • Pura-lite notifies when it is time for a new cartridge.
Specifications
Specifications
4L/min