Learn More
Hastelloy C|r gauze, 20 mesh woven from 0.23mm (0.009in) dia wire, Thermo Scientific Chemicals
High strength, corrosion resistant alloy
$36.65 - $256.65
Specifications
Chemical Name or Material | Hastelloy C™ gauze |
---|---|
Name Note | Woven from 0.23mm (0.009 in.) dia. wire |
MDL Number | MFCD00801065 |
Synonym | Nickel Molybdenum Chromium |
Solubility Information | Insoluble in water. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA40938FL
|
Thermo Scientific Chemicals
040938FL |
75 x 75 mm |
Each
for $36.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA40938GT
|
Thermo Scientific Chemicals
040938GT |
150 x 150 mm |
Each
for $84.65
Save 13% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA40938HB
|
Thermo Scientific Chemicals
040938HB |
300 x 300 mm |
Each
for $256.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Hastelloy C™ gauze is used as high strength, corrosion resistant alloy. It is used in a range of industries such as chemical processing, aerospace, pollution-control equipment, marine engineering.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hastelloy C™ gauze | |
MFCD00801065 | |
Insoluble in water. | |
Wire Cloth |
Woven from 0.23mm (0.009 in.) dia. wire | |
Nickel Molybdenum Chromium | |
20 Mesh |
Safety and Handling
GHS H Statement
H334-H351-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H351
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.