Alfa Aesar™ Hastelloy C™ gauze, 40 mesh woven from 0.19mm (0.0075 in.) dia. wire

Manufacturer: Alfa Aesar™ 46690GT

Quantity 150 x 150mm

 View more versions of this product

Catalog No. AA46690GT

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00801065
    Chemical Name or Material Hastelloy C™ gauze, 40 mesh woven from 0.19mm (0.0075 in.) dia. wire

    Specifications

    Quantity 150 x 150mm
    TSCA TSCA
    MDL Number MFCD00801065
    Mesh Size 40 Mesh
    Physical Form Wire Cloth
    Chemical Name or Material Hastelloy C™ gauze, 40 mesh woven from 0.19mm (0.0075 in.) dia. wire

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit