Learn More
BioVision IncSupplier Diversity Partner HAT Activity Fluorometric Assay Kit: Simple & convenient assay to measure HAT activity in nuclear extracts from cells and tissue & recombinant enzyme. Detection limit: < 6 mU. 100 assay DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K334100
Histone Acetyltransferases (HATs) are enzymes that acetylate histone substrates resulting in important regulatory effects on chromatin structure and assembly, and gene transcription. Modifications of these proteins by HATs play an important role in the control of gene expression, and their dysregulation has been linked to cancer, neurodegeneration, and other diseases. BioVision’s HAT Activity Assay Kit utilizes Acetyl CoA and H3 histone peptide as substrates. In this assay, HAT enzyme catalyzes the transfer of acetyl groups from Acetyl-CoA to the histone peptide, thereby generating two products - acetylated peptide and CoA-SH. The CoA-SH reacts with the developer to generate a product that is detected fluorometrically at Ex/Em = 535/587 nm. The assay can detect HAT activity as low as 6 mU in a variety of samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9700