Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HATag Rat anti-Virus, Janelia Fluor 549, Clone: 16.43, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP250416JF549

Catalog No. NBP50416549


Add to cart

Description

Description

HATag Monoclonal antibody specifically detects HATag in Virus samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

HATag
16.43
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
Rat
IgG2a
0.1mL
Primary
Virus
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
HA epitope tag, Hemagglutinin
C-terminal influenza virus hemagglutinin (HA) tag, YPYDVPDYA
Protein G purified
Store at 4°C in the dark.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit