Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAUS3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548166

Catalog No. PA548166

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Contributes to mitotic spindle assembly, maintece of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
Specifications

Specifications

HAUS3
Polyclonal
PBS with 0.09% sodium azide
Q68CZ6
HAUS augmin-like complex subunit 3, HAUS3, C4orf15
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79441
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HAUS3
Liquid
HAUS3
KLH conjugated synthetic peptide between 372-400 amino acids from the Central region of human HAUS3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit