Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAUS7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550856

Catalog No. PA550856


Add to cart

Description

Description

The antibody detects endogenous levels of total HAUS7 protein.

This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

HAUS7
Polyclonal
Unconjugated
HAUS7
Liquid
HAUS7
Synthetic peptide corresponding to residues near the N terminal of human HAUS augmin-like complex, subunit 7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q99871
UIP1, UCHL5IP
Rabbit
IgG
100 μL
-20°C
Primary
55559
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit