Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAUS8 Mouse anti-Human, DyLight 405, Clone: 15A2-35, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242904V

Catalog No. NBP242904V

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

HAUS8
15A2-35
Western Blot
Purified
Mouse
IgG1
100 ul
Primary
93323
Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
DGT4, HICE1, NY-SAR-48
The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit