Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hausser Scientific Bright-Line™ and Hy-Lite™ Counting Chambers Available on GSA/VA Contract for Federal Government customers only.

For accurate white blood cell, red blood cell, and platelet counts

Manufacturer:  HAUSSER SCIENTIFIC PARTNERSHIP 3100

 View more versions of this product

Catalog No. 02-671-10


Add to cart

Description

Description

For accurate white blood cell, red blood cell, and platelet counts

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

0.003 in.
Hausser Bright Line
Pyrex™ glass
0.1μL
Hemacytometer
0.01cm
Two 0.5mm cover glasses
Each
Counting white and red bood cells and platelets
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit