Alfa Aesar™ Haynes™ 25 gauze, 60 mesh woven from 0.14mm (0.0055 in.) dia. wire

Manufacturer: Alfa Aesar™ 46866GT

Quantity 150 x 150mm

 View more versions of this product

Catalog No. AA46866GT

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Chemical Name or Material Haynes™ 25 gauze, 60 mesh woven from 0.14mm (0.0055 in.) dia. wire

    Specifications

    Quantity 150 x 150mm
    Health Hazard 2 GHS H Statement
    H334-H372-H351-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P284a-P261-P342+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Wire Cloth
    Health Hazard 1 Danger
    Mesh Size 60 Mesh
    Chemical Name or Material Haynes™ 25 gauze, 60 mesh woven from 0.14mm (0.0055 in.) dia. wire

    GHS H Statement
    H334-H372-H351-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P284a-P261-P342+P311-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit