Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Hazardous Waste Federal Law Prohibits Improper Disposal...etc Sign
Identify containers, cylinders and drums holding hazardous waste. Brady™ Biohazard Sign help notify and warn employees about potentially dangerous materials in the facility, as required by industry regulations.
$98.00 - $173.00
Specifications
Material | Vinyl |
---|---|
Height (English) | 6 in. |
Height (Metric) | 15.2 cm |
Width (English) | 6 in. |
Width (Metric) | 15.2 cm |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
19-000-652
|
Brady™
121155 |
50/Pk. |
Pack of 50 for $98.00
|
|
|||||
19-110-971
|
Brady™
121151 |
100/Pk. |
Pack of 100 for $173.00
|
|
|||||
Description
Comply with all current Federal regulations regarding identification of hazardous waste material for trasportation off-siteSpecifications
Vinyl | |
15.2 cm | |
15.2 cm | |
Hazardous Waste Label |
6 in. | |
6 in. | |
HAZARDOUS WASTE FEDERAL LAW PROHIBITS IMPROPER DISPOSAL...etc | |
Red on Yellow |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title