Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548233

Catalog No. PA548233

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse, rat, rabbit, pig, chicken and bovine based on sequence homology.

Involved in oxygen transport from the lung to the various peripheral tissues. Spinorphin: functions as an endogenous inhibitor of enkephalin-degrading enzymes such as DPP3, and as a selective antagonist of the P2RX3 receptor which is involved in pain signaling, these properties implicate it as a regulator of pain and inflammation.
Specifications

Specifications

HBB
Polyclonal
PBS with 0.09% sodium azide
P68871
Hemoglobin subunit beta, Beta-globin, Hemoglobin beta chain, LVV-hemorphin-7, Spinorphin, HBB
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3043
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HBB
Liquid
HBB
KLH conjugated synthetic peptide between 80-107 amino acids from the C-terminal region of human HBB.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit