Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBB Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 162161AP

Catalog No. 50-173-0799


Add to cart

Description

Description

This antibody, raised against the full-length human HBB, recognizes the 13-16 kDa HBB in western blot analyses of human heart and placenta.

The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5 ft.-epsilon - gamma-G - gamma-A - delta - beta-3 ft..
Specifications

Specifications

HBB
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P68871
beta globin, CD113t C, HBB, Hemoglobin beta chain, Hemoglobin subunit beta, hemoglobin, beta
Rabbit
IgG
150 μL
-20°C
Primary
15127, 3043
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
HBB
Liquid
Hbb
HBB Fusion Protein Ag9216
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit