Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBeAg Mouse anti-Virus, DyLight 650, Clone: HBe7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB11010380C

Catalog No. NB11010380C


Add to cart

Description

Description

HBeAg Monoclonal antibody specifically detects Antigen in Virus samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications

Specifications

HBeAg
HBe7
ELISA, Sandwich ELISA
Purified
Mouse
IgG1
0.2 ml
Store at 4C in the dark.
Monoclonal
Virus
ELISA
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
HBeAg
Hybridoma clone has been derived from hybridization of Sp 2/0 myeloma cells with spleen cells of Balb/c mice immunized with recombinant HBeAg.
Protein A or G purified
RUO
Primary
Recombinant Hepatitis B virus E antigen (by ELISA).
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit