Learn More
HBQ1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 167061AP
Catalog No. 50-173-1129
Add to cart
Description
Description
Theta-globin mRNA is found in human fetal erythroid tissue but not in adult erythroid or other nonerythroid tissue. The theta-1 gene may be expressed very early in embryonic life, perhaps sometime before 5 weeks. Theta-1 is a member of the human alpha-globin gene cluster that involves five functional genes and two pseudogenes. The order of genes is: 5 ft. - zeta - pseudozeta - mu - pseudoalpha-2 -pseudoalpha-1 - alpha-2 - alpha-1 - theta-1 - 3 ft.. Research supports a transcriptionally active role for the gene and a functional role for the peptide in specific cells, possibly those of early erythroid tissue.
Specifications
Specifications
HBQ1
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09105
HBQ1, Hemoglobin subunit theta 1, Hemoglobin theta 1 chain, hemoglobin, theta 1, Theta 1 globin
Rabbit
IgG
150 μL
-20°C
Primary
3049
Immunohistochemistry (Paraffin)
Unconjugated
HBQ1
Liquid
HBQ1
HBQ1 Fusion Protein Ag10105
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human