Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBS1L Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI1C1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271628AF405

Catalog No. N271628A405


Add to cart

Description

Description

HBS1L Monoclonal antibody specifically detects HBS1L in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

HBS1L
OTI1C1
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
HBS1L
Recombinant protein fragment corresponding to amino acids 1-260 of human HBS1L (NP_006611) produced in E.coli.
Immunogen affinity purified
Signal Transduction
Primary
10767.0
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp434g247, ERF3-similar protein, ERFSDKFZp686L13262, HBS1 (S. cerevisiae)-like, HBS1KIAA1038EF-1a, HBS1-like (S. cerevisiae), HBS1-like protein, Hsp70 subfamily B suppressor 1-like protein, HSPC276
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit