Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBsAg Antibody (1044/329), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064554F
Description
HBsAg Monoclonal antibody specifically detects HBsAg in Virus samples. It is validated for ELISA, ImmunofluorescenceSpecifications
HBsAg | |
Monoclonal | |
FITC | |
HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
Native | |
0.1 mL | |
Cell Biology, Virology Bacteria and Parasites | |
944569 | |
Store at 4°C in the dark. | |
IgG1 |
ELISA, Immunofluorescence | |
1044/329 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction