Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HBsAg Goat anti-Virus, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
$176.85 - $436.00
Specifications
Antigen | HBsAg |
---|---|
Concentration | 4.0 mg/mL |
Dilution | ELISA 1:200 - 1:1000 |
Applications | ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10062647
|
Novus Biologicals
NB10062647 |
0.5 mL |
Each for $436.00
|
|
NB017502
|
Novus Biologicals
NB100626470.1ML |
0.1 mL |
Each for $176.85
|
|
Description
HBsAg Polyclonal specifically detects HBsAg in Virus samples. It is validated for ELISA.Specifications
HBsAg | |
ELISA 1:200 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Phosphate Buffered Saline with 0.1% Sodium Azide | |
944569 | |
Hepatitis B, subtypes ad and ay | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
4.0 mg/mL | |
ELISA | |
Unconjugated | |
Goat | |
Virus | |
HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
S | |
IgG | |
IgG purified | |
NB100-62647 is specific for Hepatitis B surface antigen (HBsAg), a protein component of the viral envelope which is predominantly expressed in the cytoplasm of infected cells. Hep B is a major causative agent of acute and chronic liver disease in humans. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title