Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBsAg Mouse anti-Human, Virus, Clone: 30000000, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen HBsAg
Clone 30000000
Classification Monoclonal
Conjugate Unconjugated
Form Supernatant
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP122568
View Documents
Novus Biologicals
NBP122568
0.5 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

HBsAg Monoclonal antibody specifically detects HBsAg in Human, Virus samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

HBsAg
Monoclonal
Supernatant
RUO
Tissue culture supernatant, containing less than 0.1% sodium azide with Sodium Azide
944569
Purified HBsAg subtype ayw3.
Primary
Store at 4°C.
30000000
Unconjugated
Mouse
Virology Bacteria and Parasites
HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
S
IgG1
Tissue culture supernatant
This reacts with the 'a' determinant present on the HBsAg subtypes ayw1, ayw2, ayw3, ayw4, ayr, adw2, adw4 and adr. It does not react with normal tissues.
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit