Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBsAg Mouse anti-Virus, Janelia Fluor 646, Clone: 1044/329, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB10064554JF646

Catalog No. NB064554646


Add to cart

Description

Description

HBsAg Monoclonal antibody specifically detects HBsAg in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

HBsAg
1044/329
Immunocytochemistry/Immunofluorescence
Purified
S
Native
Protein A purified
Cell Biology, Virology Bacteria and Parasites
Primary
944569
Virus
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Recognizes the Hepatitis B surface antigen; in a simple ELISA using Biogenesis Test Groups (BTGs) this is only reactive with Hepatitis B surface antigen.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit