Learn More
HBXIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 144921AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-172-9488
Add to cart
Description
Description
HBXIP, also named as XIP, belongs to the HBXIP family. It is originally identified by its interaction with the C-terminus of hepatitis B virus (HBV) X protein (HBx). HBXIP is a regulator of centrosome dynamics and cytokinesis. HBXIP is able to up-regulate S100A4 though activating STAT4 and inducing DNA methylation of PTEN. HBXIP is a cellular 18kd protein and cytoplasm stain.
Specifications
Specifications
HBXIP
Polyclonal
Unconjugated
LAMTOR5
HBV X interacting protein, HBX interacting protein, HBXIP, XIP
Rabbit
Antigen Affinity Chromatography
RUO
10542, 295357, 68576
-20°C
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43504, Q9D1L9
LAMTOR5
HBXIP Fusion Protein Ag5894
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG