Learn More
HCA66 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | HCA66 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2598
![]() |
Bethyl Laboratories, Inc
IHC00719T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2597
![]() |
Bethyl Laboratories, Inc
IHC00719 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Northern white-cheeked gibbon, Crab-eating macaque and White-tufted-e
UTP6 involved in nucleolar processing of pre-18S ribosomal RNA.Specifications
HCA66 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
C17orf40, HCA66, U3 small nucleolar RNA-associated protein 6 homolog, hepatocellular carcinoma associated antigen 66, hepatocellular carcinoma-associated antigen 66, multiple hat domains protein | |
UTP6 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q9NYH9 | |
55813 | |
Between 565 and 597 | |
Primary | |
4° C | |
UTP6 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.