Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACY3 Mouse anti-Human, Clone: OTI2G4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525389

Catalog No. PIMA525389


Add to cart

Description

Description

ACY3 belongs to the aspA/astE family. Aspartoacylase subfamily. Catalytic activity: N-acyl-L-aspartate + H(2)O = a carboxylate + L-aspartate.
Specifications

Specifications

ACY3
Monoclonal
0.42 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96HD9
N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); Acylase III; Aminoacylase-3; ACY-3; Aspartoacylase-2; Hepatitis C virus core-binding protein 1; HCBP1; HCV core-binding protein 1; ACY3; ASPA2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
91703
Flow Cytometry, Western Blot
OTI2G4
Unconjugated
ACY3
Liquid
ACY3
Full length human recombit protein of ACY3 produced in 293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit