Learn More
Sigma Aldrich Fine Chemicals Biosciences HCC827 GR6 Human Female Lung Adenocarcinoma Metastasis Cell Line with EGFR Exon 19 Deletion | 1 Vial
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SCC426
Encompass
The HCC827 GR6 cell line was derived from a human female lung adenocarcinoma metastasis. This adherent epithelial cell line has a deletion in exon 19 of the EGFR gene, which increases its sensitivity to EGFR tyrosine kinase inhibitors including gefitinib and erlotinib.
Catalog No. NC2372274
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.