Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HCFC1 Polyclonal Antibody

Rabbit Polyclonal Antibody

$209.50 - $511.50

Specifications

Antigen HCFC1
Concentration 0.81 μg/mL
Content And Storage -20°C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119198
View Documents
Invitrogen™
PA5119198
200 μL
Each for $511.50
Only null left
Add to Cart
 
PIPA5121921
View Documents
Invitrogen™
PA5121921
20 μL
Each for $209.50
Only null left
Add to Cart
 
Description

Description

HCFC1 Polyclonal Antibody for Western Blot, IHC (P)

Tumor necrosis factor (TNF) receptor associated factors (TRAFs) were initially discovered as adaptor proteins that link the TNF receptor superfamily to signaling pathways and are thus important regulators of cell death and cellular response to stress. TRAF proteins share a homology region that allows them to bind to cell receptors and other TRAF proteins, causing the activation of different signal cascades depending on the TRAFs involved. For example, TRAF2 and TRAF3 directly bind to the CD40, a NF receptor superfamily member involved in inducing B cell immunity, and are critical for NF-kappa-B activation in mouse B lymphocytes. TRAF2 along with TRAF6 has also been shown to be required for CD40 signaling in nonhemopoietic cells. TRAF2 also interacts with the TRFR superfamily member lymphotoxin-beta receptor (LTbetaR) in association with TRAF3 and the apoptosis inhibitors cIAP1 and Smac.
Specifications

Specifications

HCFC1
-20°C
Polyclonal
Liquid
IgG
Human
HCFC1
AW060991; C1 factor; CFF; HCF; HCF C-terminal chain 1; HCF C-terminal chain 2; HCF C-terminal chain 3; HCF C-terminal chain 4; HCF C-terminal chain 5; HCF C-terminal chain 6; HCF N-terminal chain 1; HCF N-terminal chain 2; HCF N-terminal chain 3; HCF N-terminal chain 4; HCF N-terminal chain 5; HCF N-terminal chain 6; HCF1; HCF-1; hcfc1; HFC1; host cell factor 1; host cell factor C1; host cell factor C1 (VP16-accessory protein); LOW QUALITY PROTEIN: host cell factor 1; MRX3; PPP1R89; protein phosphatase 1, regulatory subunit 89; RGD1563804; VCAF; VP16 accessory protein; VP16-accessory protein
HCFC1
Primary
Antigen affinity chromatography
0.81 μg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300
P51610
3054
Recombinant protein Host Cell Factor C1. The antigen corresponds to amino acid range 52-258 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.