Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCFC1R1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548252

Catalog No. PA548252

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with chicken and mouse based on sequence homology.

Regulates HCFC1 activity by modulating its subcellular localization. Overexpression of HCFC1R1 leads to accumulation of HCFC1 in the cytoplasm. HCFC1R1-mediated export may provide the pool of cytoplasmic HCFC1 required for import of virion-derived VP16 into the nucleus.
Specifications

Specifications

HCFC1R1
Polyclonal
PBS with 0.09% sodium azide
Q9NWW0
Host cell factor C1 regulator 1, HCF-1 beta-propeller-interacting protein, HCFC1R1, HPIP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54985
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HCFC1R1
Liquid
HCFC1R1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human HCFC1R1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit