Learn More
Aviva Systems Biology Corp HCG_1660138 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP50998_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against hCG_1660138. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human hCG_1660138. Alternate names: LOC139542, hCG_1660138ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60481
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.