Learn More
Aviva Systems Biology Corp HCG_1982709 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP51031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against hCG_1982709. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human hCG_1982709. Alternate names: LOC441488, hCG_1982709ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60489
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.