Learn More
Aviva Systems Biology Corp hCG_20426 Antibody - N-terminal region (ARP47801_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47801_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against hCG_20426. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human hCG_20426. Alternate names: LOC441869, hCG_20426ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-59667