Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hCG beta-7 Monoclonal Antibody (6F12), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524507
Description
Peptide Sequence: GYCPTMTRVL QGVLPALPQV VCNYRDVRFE SIRLPGCPRG VNPVVSYAVA LSCQCALCRR STTDCGGPKD HPLTCDDPRF QASSSSKAPP PSLPSPSRLP GPSDTPILPQ
hCG beta-7 Monoclonal antibody specifically detects hCG beta-7 in Human samples. It is validated for ELISA, Western Blot.Specifications
hCG beta-7 | |
Monoclonal | |
Unconjugated | |
CGB7 | |
CGB6; CGB7; CG-beta-a; Choriogonadotropin subunit beta 7; chorionic gonadotropin beta 7 subunit; chorionic gonadotropin beta subunit 7; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide 7; human chorionic gonadotropin | |
Mouse | |
Protein A | |
RUO | |
94027 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
6F12 | |
PBS with no preservative; pH 7.4 | |
P0DN87 | |
CGB7 | |
CGB7 (NP_149133, 56 a.a. ∽ 165 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction