Learn More
HCG Mouse anti-Human, Clone: CG04 + CG05, Abnova™
Mouse monoclonal antibody raised against human hCG.
Manufacturer:  Abnova Corporation MAB12738
Catalog No. 89-934-230
Add to cart
Description
Description
Specifications
Specifications
HCG
CG04 + CG05
Mouse monoclonal antibody raised against human hCG.
In 10mM PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Mouse
IgG2a, kappa; IgG1, kappa
250 uL
Store at 4°C. Do not freeze.
Monoclonal
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Purified Beta-subunit of HCG.
Protein A/G purification
RUO
Primary
Human