Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chorionic Gonadotropin alpha Chain (hCG alpha) Mouse anti-Human, Mouse, Clone: AH33-20, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267281

Catalog No. NBP267281


Add to cart

Description

Description

Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal antibody specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Chorionic Gonadotropin alpha Chain (hCG alpha)
AH33-20
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.2% Sodium Azide
CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA
Mouse
IgG1
100μL
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
Western Blot 1:1000, ELISA 1:5000-1:10000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Purified
CGA
Full length protein
Protein A purified
Cancer
Primary
1081
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit