Learn More
HCLS1 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB30128
Catalog No. 89-033-416
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human HCLS1.
  • High specificity
  • High reactivity
Specifications
Specifications
HCLS1
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (1.25 ug/mL) The optimal working dilution should be determined by the end user.
NM_005335
CTTNL|HS1
Rabbit
Affinity purification
RUO
Primary
3059
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human HCLS1.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
HCLS1
A synthetic peptide corresponding to N-terminus of human HCLS1.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)