Learn More
HCN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00122W
Catalog No. PIOSR00122W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
Hyperpolarization-activated ion channel exhibiting weak selectivity for potassium over sodium ions. Contributes to the native pacemaker currents in heart (If) and in neurons (Ih). Activated by cAMP, and at 10-100 times higher concentrations, also by cGMP. May mediate responses to sour stimuli. The potassium channel is probably composed of a homo- or heterotetrameric complex of pore-forming subunits. Heteromultimer with HCN2. Interacts with KCNE2. Detected in brain, in particular in amygdala and hippocampus, while expression in caudate nucleus, corpus callosum, substantia nigra, subthalamic nucleus and thalamus is very low or not detectable. Detected at very low levels in muscle and pancreas. The segment S4 is probably the voltage-sensor and is characterized by a series of positively charged amino acids at every third position.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HCN1
Polyclonal
HCN1
Brain cyclic nucleotide-gated channel 1, BCNG-1, BCNG1, HAC-2
Rabbit
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
O60741, O88704, Q9JKB0
HCN1
A synthetic peptide from cytoplasmic region of human HCN1 conjugated to an immunogenic carrier protein
RUO
15165, 348980, 84390
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations