Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTLA-4 Human anti-Human, Alexa Fluor 350, Clone: Hu111, R&D Systems
Human Monoclonal Antibody
Manufacturer: R&D Systems FAB9928U100UG
This item is not returnable.
View return policy
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry.Specifications
CTLA-4 | |
Hu111 | |
Flow Cytometry 0.25-1 μg/10^6 cells | |
Conjugated, Purified, Supernatant | |
Human | |
IgG1 | |
100 μg | |
Primary | |
1493 | |
Human |
Flow Cytometry | |
Alexa Fluor 350 | |
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
Human CTLA-4 | |
Protein A or G purified from cell culture supernatant | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
Monoclonal | |
Detects Human CTLA-4 based on Ipilimumab therapeutic antibody.This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Ipilimumab. This product is for research use only. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title