Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCV E2 Human anti-Virus, Clone: AR3A, MilliporeSigma™

Human Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen HCV E2
Clone AR3A
Concentration 0.5 mg/mL
Applications ELISA, Neutralization
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2336100
View Documents
MilliporeSigma
MABF2336100UL
100 μL
Each for $373.00
Add to cart
 
MABF233625
View Documents
MilliporeSigma
MABF233625UL
25 μL
Each for $145.80
Add to cart
 
Description

Description

HCV E2 Monoclonal antibody specifically detects HCV E2 in Virus samples. It is validated for ELISA, Neutralizing.
Specifications

Specifications

HCV E2
0.5 mg/mL
Unconjugated
Human
Virus
Antibody Fab fragment C1 expressed in E. coli, purified, and converted into full-length IgG1 molecule in CHO-K1 cells.
Monoclonal
Protein G purified
AR3A
ELISA, Neutralization
Purified
RUO
Purified human recombinant recombinant monoclonal antibody in PBS without preservatives.
IgG1
Primary
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit