Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HD domain containing 2 Rabbit anti-Human, Alexa Fluor 594, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297983AF594
Description
HD domain containing 2 Polyclonal specifically detects HD domain containing 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HD domain containing 2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C6orf74, CGI-130, chromosome 6 open reading frame 74, dJ167O5.2, HCV NS5A-transactivated protein 2, HD domain containing 2, HD domain-containing protein 2, Hepatitis C virus NS5A-transactivated protein 2, MGC87330, NS5ATP2 | |
Produced in rabbits immunized with E. coli-derived Human HD domain containing 2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
51020 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title