Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HD-polyAla Ct Rabbit anti-Human,Mouse, Unconjugated, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$153.50 - $386.00

Specifications

Antigen HD-polyAla Ct
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Serum
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ABN167210MI
View Documents
MilliporeSigma
ABN1672100UL
100 μL
Each for $386.00
Add to cart
 
ABN167225MI
View Documents
MilliporeSigma
ABN167225UL
25 μL
Each for $153.50
Add to cart
 
Description

Description

HD-polyAla Ct Polyclonal antibody specifically detects HD-polyAla Ct in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.
Specifications

Specifications

HD-polyAla Ct
Unconjugated
Rabbit
HD-RAN-PolyAla;HD-PolyAla
Polyclonal
Unpurified
Immunocytochemistry, Immunohistochemistry, Western Blot
Serum
Human, Mouse
A synthetic peptides from the C-terminal regions of the predicted 21 polyAla frame of HD in the CAG direction.
Primary
Stable for 1 year at -20°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit