Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HD-polyLeu-Ct Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$184.00 - $463.00
Specifications
Antigen | HD-polyLeu-Ct |
---|---|
Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Serum |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN167025MI
|
MilliporeSigma
ABN167025UL |
25 μL |
Each for $184.00
|
|
|||||
ABN167010MI
|
MilliporeSigma
ABN1670100UL |
100 μL |
Each for $463.00
|
|
|||||
Description
HD-polyLeu-Ct Polyclonal antibody specifically detects HD-polyLeu-Ct in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.Specifications
HD-polyLeu-Ct | |
Polyclonal | |
Serum | |
Neuroscience | |
HD-RAN PolyLeu;HD-PolyLeu | |
IgG | |
Unpurified | |
This rabbit polyclonal antibody specifically detects RAN proteins with PolyLeu-HD. |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
A synthetic peptide corresponding to the C-terminal region of the predicted polyLeu frames of HD in the CTG direction (HD-Leu-Ct). | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
∼21 kDa observed. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title