Learn More
Aviva Systems Biology Corp HDAC1 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP32358_P050
This is a rabbit polyclonal antibody against HDAC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HDAC1. Alternate names: DKFZp686H12203, GON-10, HD1, RPD3, RPD3L1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54542