Learn More
HDAC1 Mouse anti-Human, Clone: C.15100144, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259238
Catalog No. NBP259238
Add to cart
Description
Description
HDAC1 Monoclonal antibody specifically detects HDAC1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Knockdown Validated.
Specifications
Specifications
HDAC1
C.15100144
Western Blot 1:1000, Chromatin Immunoprecipitation 3 ul
Supernatant
HDAC1
The exact immunogen is propietary information.
Concentrated Supernatant
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, ChIP assay
Unconjugated
Culture Medium with 0.05% Sodium Azide
EC 3.5.1.98, GON-10, HD1DKFZp686H12203, histone deacetylase 1, reduced potassium dependency, yeast homolog-like 1, RPD3L1RPD3
Mouse
IgG1
100 ul
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Histone Deacetylases, Loading Controls, Stem Cell Markers
Primary
3065