Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC1 Monoclonal Antibody (10E2), Active Motif™
Mouse Monoclonal Antibody
$147.50 - $576.30
Specifications
Antigen | HDAC1 |
---|---|
Clone | 10E2 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-4035
|
Active Motif
39532 |
10 μL |
Each for $147.50
|
|
|||||
50-199-4034
|
Active Motif
39531 |
100 μL |
Each for $576.30
|
|
|||||
Description
Histone deacetylase (HDAC) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones - H2A, H2B, H3, and H4. Histone deacetylation gives a tag for epigenetic repression and plays and important role in transcriptional regulation, cell cycle progression, and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.Specifications
HDAC1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
antibody with 30% glycerol and 0.035% sodium azide; pH | |
DKFZp686H12203; GON-10; HD1; HDAC1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1 | |
HDAC1 | |
IgG1 | |
-20°C, Avoid Freeze/Thaw Cycles | |
HDAC1 |
10E2 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
O09106, Q13547, Q4QQW4 | |
297893, 3065, 433759 | |
This HDAC1 antibody was raised against a kLH-conjugated peptide corresponding to amino acids 467-482 of human and murine HDAC1. | |
Primary | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title