Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC10 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600223IR
Description
HDAC10 Polyclonal antibody specifically detects HDAC10 in Human samples. It is validated for Western BlotSpecifications
HDAC10 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 755 | |
DKFZP761B039, EC 3.5.1.98, HD10, histone deacetylase 10, MGC149722 | |
Synthetic peptide corresponding to a portion of human HDAC 10. | |
0.1 mL | |
Breast Cancer, Cancer, Checkpoint signaling, Chromatin Research, Epigenetics, Transcription Factors and Regulators | |
83933 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction