Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC10 Mouse anti-Human, Clone: OTI4D4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA500752

Catalog No. 50-167-1893


Add to cart

Description

Description

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.
Specifications

Specifications

HDAC10
Monoclonal
0.68 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q969S8
HD10
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
83933
Flow Cytometry, Immunoprecipitation, Western Blot
OTI4D4
Unconjugated
HDAC10
Liquid
HDAC10
Full length human recombit protein of human HDAC10 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit